Learn More
Abnova™ CLDN18 (Texas Red/FITC) Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FS0037.100uL
Additional Details : Weight : 0.02000kg
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Liquid | |
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique. | |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
Approx. 430kb/720kb | |
100 μL | |
Human |
Texas Red, FITC | |
Fluorescent In Situ Hybridization (Cell) | |
3q23/3q23 | |
CLDN18 Split FISH Probe | |
Genomic DNA |