missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Clear Borosilicate Glass Volumetric Flasks

Conduct experiments in the laboratory with Fisherbrand™ Clear Borosilicate Glass Volumetric Flasks. They are graduated for precise measurement.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10024212

  • 62.32 € / Pack of 5


In stock, ready to ship: 4
Add to basket

You may only order up to the available stock. For more details call customer services on 01 885 5854 .

Description

Description

  • Graduated
  • Polyethylene snap cap
  • Clear
Specifications

Specifications

49 mm
Clear
Borosilicate Glass
5 Pack
ISO 1042, DIN 12664 Pt I, BS1792
50 mL
139 mm
±0.06 mL
Videos
SDS
Documents