missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ EdU Colorimetric IHC Detection Kit

Invitrogen™ Click-iT™ EdU Colorimetric IHC Detection Kit is a superior alternative to traditional methods for the colorimetric detection of proliferating cells within tissue sections.

Brand:  Invitrogen™ C10644

Additional Details : Weight : 140,050.00000kg

Product Code. 15642436

  • 768.00 € / Pack of 50
In Stock
to see stock.
Add to basket

Description

Description

  • Substrate: DAB
  • Substrate Type: HRP
  • The Click-iT™ EdU Colorimetric IHC (Immunohistochemistry) Detection Kit is a superior alternative to traditional methods for the colorimetric detection of proliferating cells within tissue sections. In this assay, 5-ethynyl-2'-deoxyuridine (EdU), a nucleoside analog of thymidine containing an alkyne moiety, is incorporated into newly synthesized DNA. When biotin containing an azide group is added, a brief click reaction attaches the azide to the alkyne moiety on the EdU molecule. Upon addition of streptavidin–peroxidase and a peroxidase substrate, an enzymatic reaction generates a signal that can be detected with light microscopy and stored for future analysis.
  • Optimized - enables colorimetric detection of proliferating cells in either tissue or cell samples
  • Efficient - no denaturation steps or harsh treatment required
  • Robust - improved preservation of cell morphology and the cellular environment offers content-rich results
  • Consistent - not dependent on variable antibody lots for detection
  • Simple - designed to work consistently, in less time than traditional methods
Specifications

Specifications

HRP
EVOS™ XL Core Imaging System, EVOS™ XL Imaging System
Store at 2°C to 8°C.
DAB
Click-iT™
IHC Detection Kit
Colorimetric
Slides
Wet Ice
HRP
IHC (Immunohistochemistry)
All species
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.