missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) iodide, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049816
Click to view available options
Quantity:
25 g
100 g
15238-00-3
MFCD00016030
cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis
15238-00-3
MFCD00016030
cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis

Moisture and humidity indicator, catalyst

Cobalt(II) iodide is used as a moisture and humidity indicator. It is also used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 570°C (vac.)
Melting Point 515°C (vac.)
Physical Form Powder
Quantity 25 g
Merck Index 14,2443
Solubility Information Soluble in water
Formula Weight 312.74
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Assay (metals basis)
Chemical Name or Material Cobalt(II) iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'