missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(III) fluoride, 99%, Thermo Scientific Chemicals

Catalog Number p-4407410
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
10026-18-3
CoF3
MFCD00010942
This item is not returnable. View return policy
10026-18-3
CoF3
MFCD00010942

Cobalt(III) fluoride is a powerful fluorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
Formula Weight 115.93
Odor Acrid
Sensitivity Moisture sensitive
Appearance Light brown powder
Density 3.88
Chemical Name or Material Cobalt(III) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'