missing translation for 'onlineSavingsMsg'
Learn More

Cobalt oxide, typically 3.4-4.5%, Molybdenum oxide typically 11.5-14.5% on alumina, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 045579.36

Code : NEW

Additional Details : Weight : 0.50000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

 View more versions of this product

Product Code. 15424005

  • 420.00 € / 500g
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cobalt oxide, typically 3.4-4.5%, Molybdenum oxide typically 11.5-14.5% on alumina is used in the preparation of biofuel production by using algae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt oxide, Molybdenum oxide
Blue
500 g
UN3077
Co oxide:typically 3.4 to 4.5%; Mo oxide: typically 11.5 to 14.5%
On alumina
Trilobe Extrudate
MFCD01074445
Insoluble in water.
Odorless
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only