missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium bromide, 99.9%, pure, Thermo Scientific Chemicals
86.80 € - 220.00 €
Chemical Identifiers
CAS | 7787-69-1 |
---|---|
Molecular Formula | BrCs |
Molecular Weight (g/mol) | 212.81 |
MDL Number | MFCD00010954 |
InChI Key | LYQFWZFBNBDLEO-UHFFFAOYSA-M |
Synonym | cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide |
PubChem CID | 24592 |
IUPAC Name | cesium;bromide |
SMILES | [Br-].[Cs+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10437780
|
Thermo Scientific Acros
192060250 |
25 g | Glass bottle |
86.80 €
25g |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
View alternative products » | |||||||||
10447780
|
Thermo Scientific Acros
192061000 |
100 g | Glass bottle |
220.00 €
100g |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
View alternative products » | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-69-1 | |
212.81 | |
LYQFWZFBNBDLEO-UHFFFAOYSA-M | |
24592 | |
[Br-].[Cs+] |
BrCs | |
MFCD00010954 | |
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide | |
cesium;bromide |
Specifications
1300.0°C | |
636.0°C | |
7787-69-1 | |
Crystalline Powder and/or Crystals | |
99.90% | |
CsBr | |
15, 2008 | |
Solubility in water: 55g/100mL (20 c). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone | |
[Br-].[Cs+] | |
212.81 | |
212.81 | |
Pure | |
4.44 |
Cesium bromide | |
99.9% | |
White | |
25 g | |
BrCs | |
MFCD00010954 | |
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide | |
LYQFWZFBNBDLEO-UHFFFAOYSA-M | |
cesium;bromide | |
24592 | |
99.9% | |
Glass bottle | |
4.4400g/mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cesium bromide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only