missing translation for 'onlineSavingsMsg'
Learn More

SCAT Collecting Tray, HDPE-EC

SCAT Collecting Trays store your waste containers and provide additional safety in case of any leakage.

Brand:  SCAT 117984

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10282710

  • 111.00 € / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

  • Suitable for a wide range of canisters
  • Collect liquid in case of a leakage
  • Available with base insert so the canisters stay always clean and dry
  • Optional with electrostatic conductive material
  • Made of PE-HD or optional PE-HD-ec
Specifications

Specifications

23 L
Collecting tray, PE-HD, dimensions (W x H x D): 290 x 200 x 385 mm (interior), dimensions (W x H x D): 340 x 210 x 465 mm (exterior)
210 mm
HDPE
340 mm
Handling of Hazardous Substances
Rectangular
Inner: 290 x 200 x 385 mm, Outer: 340 x 210 x 465 mm
White
465 mm
Collection Tray
Waste Containers
1 Ea.
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers