Learn More
Buffer Solution, pH 10.00, Blue, Reagecon™
Certified by an ISO 17025 accredited method and NIST traceable. Reagecon™ Buffer Solution, pH 10.00, Blue is tested at 20°C to an accuracy of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, is color coded and features a twin-neck bottle design.
Brand: Reagecon 11005CTT
Code : NEW
Additional Details : Weight : 0.52020kg
Description
Color coded and packaged for convenience
- Colored yellow for easy identification of pH value
- Supplied in a Twin Neck bottle that features an integrated calibrating chamber
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
0°C | |
Blue | |
Liquid | |
Colored pH Buffer | |
Twin Neck bottle with integrated calibrating chamber | |
Fully miscible in water |
100°C | |
10.00 ±0.01 | |
500 mL | |
Characteristic | |
Yes |
Safety and Handling
- BUFFER SOLUTION pH 10.00 ± 0.01 @ 20°C
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Boric Acid, DisodiumSalt