missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Upright Freezers, 130 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
167.00 € - 217.00 €
Specifications
Height (Metric) | 130 mm (Box) |
---|---|
For Use With (Equipment) | Upright Freezers |
Material | Stainless Steel |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15250864
|
Tenak™
TE25202 |
Boxes (8) |
217.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15230864
|
Tenak™
TE25200 |
Boxes (6) |
167.00 €
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15260864
|
Tenak™
TE25204 |
Boxes (12) |
191.00 €
Each |
In Stock
Log in to see stock. |
|||||
15280864
|
Tenak™
TE25208 |
Boxes (15) |
203.00 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
Specifications
130 mm (Box) | |
Upright Freezers | |
Stainless Steel |