Learn More
Fisherbrand™ Compact Balance
The perfect economical portable balance, ideal when cost matters and dependable results are essential. Fisherbrand™ Compact Balance offers capacities of 200g to 5kg and readability of 0.1g or 1g for use classroom and educational settings.
41.20 € - 59.30 €
Specifications
Electrical Requirements 3 AA Batteries (included) or optional AC adapter
Display Type LCD
Stabilization Time 3 sec.
View More Specs
Products 4
Product Code Brand Capacity (Metric) Readability Price Quantity & Availability  
Product Code Brand Capacity (Metric) Readability Price Quantity & Availability  
15335103 View accessory products View Documents Fisherbrand™
5,000g 1 g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.
N/A View alternative products
Save Up to 
»
15305103 View accessory products View Documents Fisherbrand™
200g 0.1 g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.
N/A View alternative products
Save Up to 
»
15315103 View accessory products View Documents Fisherbrand™
500g 0.1 g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.
N/A View alternative products
Save Up to 
»
15325103 View accessory products View Documents Fisherbrand™
2,000g 1 g This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
  • Sleek, low profile design
  • Large, high contrast LCD
  • Energy saving auto shut-off feature
  • Two-button keypad
  • Storage made easy with a stackable design
  • Integrated load cell lock
  • Overload and under-load indicators
  • Digital calibration from keypad
  • Low battery indicator
  • Three AA batteries (included) or optional a. c. adapter
  • Two year warranty
Specifications
Specifications
3 AA Batteries (included) or optional AC adapter
LCD
3 sec.