missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Conical Shape PTFE Key Separating Funnels

Ideally suited for phase separation

Brand:  Duran™ 1064807

Code : A0

Additional Details : Weight : 0.26400kg

 View more versions of this product

Product Code. 10426332

  • 94.15 € / Each


Estimated Shipment
15-03-2019
Add to basket

Description

Description

  • Conical shape, with PTFE key
  • Includes standard ground stopcock, retaining device and plastic stopper
Specifications

Specifications

10mm
500mL
Borosilicate Glass
Separating funnel
320mm
Videos