Learn More
Emma Safety Footwear Constans Safety Shoes
High quality full grain leather upper and nappa leather lining. Exclusively developed for semi-orthopedic adjustments especially where customized in-soles are required.
Brand:  Emma Safety Footwear MM13207045
Additional Details : Weight : 2.30100kg
 View more versions of this product
Product Code. 15948221
  • 381.60 € / 1 pair
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Vibram™ Nitrile Rubber Heat Resistant Outsole
Specifications
Specifications
EN ISO 20345:2011
Black
Full grain Nappa leather
45
Laces
Constans
Leather
Aluminum