Learn More
Contec™ QuickTask™ Mop
The Contec™ QuickTask™ Flat Mops are ideally suited for cleaning and applying disinfectant to ceilings, walls and floors including perforated cleanroom floors and grates. All QuickTask hardware is fully autoclavable.
Brand: Contec™ TASK0250
220.00 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Additional Details : Weight : 3.60000kg
Description
Contec's QuickTask product line consists of a variety of knitted polyester, polyester/cellulose, microfiber/foam, and microfiber flat mop heads that are easily installed onto a hinged mop head frame and handle assembly.
Each mop head has been specifically chosen to be compatible with a wide range of chemicals and disinfectants including quats and hypochlorite solutions. QuickTask mop head frames are available in stainless steel or durable autoclavable plastic.
The lightest mop cover in the range, the sorbent cleanroom laundered multi-layer knitted polyester is ideal for use on smooth surfaces. Its sorbent knit structure applies solutions evenly and removes contaminants uniformly. Available validated sterile by gamma irradiation.
Specifications
16 x 5 in. (41 x 13 cm) | |
Yes | |
No |
Sterile | |
2906, 2699P, 2740 | |
Knitted Polyester Fabric |