missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Converter Plate Uv 302Nm To Uv 365Nm
Brand: UVP 38-0325-01
Code : 04
Additional Details : Weight : 1.70000kg
Brand: UVP 38-0325-01
Code : 04
Additional Details : Weight : 1.70000kg