Learn More
Corning™ CoolBox™ XT Systems
Provide sample cooling or freezing without ice, electricity, or batteries.
Marque: Corning™ 432030
Code : NEW
Informations supplémentaires : Weight : 4.00000kg
Description
“Limited quantities remain. Buy now at a deep discount.”
(Discontinued: 432030; Suggested Replacement: 432022 + 432040)
Ready to Use MicroTube 24 CoolBox XT System comes complete with CoolBox, cooling core and M24 CoolRack module.
- For use with 24 x 1.5 mL or 2.0 mL microfuge tubes
- Includes: CoolBox XT System, SBS-compatible CoolRack XT M24
- Suggested application: Enzyme, PCR reagent or other biological specimen cooling
Spécification
Benchtop cooling system | |
Cryo Storage Box | |
24 (1.5 mL or 2 mL Cryogenic Vials) | |
CoolBox XT M24 system |
Green | |
Enables Cooling for 16 Hours, Freezing for 8 Hours | |
CoolBox XT System, SBS-Compatible CoolRack XT M24 | |
4 kg |