Learn More
Corning™ CoolBox™ XT Systems
Provide sample cooling or freezing without ice, electricity, or batteries.
Brand: Corning™ 432032
Code : NEW
Additional Details : Weight : 4.00000kg
Description
'Limited quantities remain. Buy now at a deep discount.'
(Discontinued: 432032; Suggested Replacement: 432022 + 432054)
Ready to Use PCR Strip CoolBox XT System CoolBox System comes complete with CoolBox, cooling core and PCR CoolRack module.
- For use with 6 strip wells and 12 microfuge tubes (1.5 to 2.0 mL)
- Includes: CoolBox XT System, SBS-compatible CoolRack XT M-PCR
- Suggested application: PCR/qPCR strip well and reagent tube preparation
Specifications
Benchtop cooling system | |
Cool Box | |
12 (1.5 to 2 mL Microfuge Tubes) | |
CoolBox XT PCR Strip system |
Green | |
Enables Cooling for 16 Hours, Freezing for 8 Hours | |
CoolBox XT System, SBS-Compatible CoolRack XT M-PCR | |
4 kg |