Learn More
Corning™ CoolBox™ XT Systems
Provide sample cooling or freezing without ice, electricity, or batteries.
Brand: Corning™ 432033
Code : NEW
Additional Details : Weight : 7.00000kg
Description
“Limited quantities remain. Buy now at a deep discount.”
(Discontinued: 432033; Suggested Replacement: 432026 + 432053 + 432040)
Ready to Use PCR Sample Preparation CoolBox 2XT System comes complete with CoolBox, cooling core and both M24 and PCR96 CoolRack module.
- For use with one 96-well PCR plate or strip wells or tubes and 24 x 1.5 mL or 2.0 mL microfuge tubes
- Includes: CoolBox 2XT System, SBS-compatible CoolRack XT M24, CoolRack PCR96
- Suggested application: PCR/qPCR plate and reagent tube preparation
Specifications
Benchtop cooling system | |
Cool Box | |
96 (1.5 to 2 mL Microfuge Tubes) | |
CoolBox 2XT PCR system |
Green | |
Enables Cooling for 16 Hours, Freezing for 8 Hours | |
CoolBox 2XT System, CoolRack PCR96, SBS-Compatible CoolRack XT M24 | |
7 kg |