missing translation for 'onlineSavingsMsg'
Learn More

Copper, AAS standard solution, Specpure™ Cu 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4408544
Click to view available options
Quantity:
100 mL
500 mL
7440-50-8, 7732-18-5
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
CHEBI:30052
copper
[Cu]
7440-50-8, 7732-18-5
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
CHEBI:30052
copper
[Cu]

Copper, AAS standard solution is used to determine copper in samples of water and water-suspendedsediment with a specific conductance not greater than 10,000 μS/cm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mL
Color Colorless
Grade AAS Standard
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Copper: 0.1%; Water: 94.9%
Vapor Pressure 17mm Hg at 20°C
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Copper

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'