missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated, 99.8% (metals basis)

For Graphene growth

Brand:  Alfa Aesar™ 046365.RH

Code : NEW

Additional Details : CAS Number : 7440-50-8 Weight : 0.01000kg

Quantity 30 x 1000cm

 View more versions of this product

Product Code. 15475015

  • 180.00 € / Each


Estimated Shipment date
from Supplier 23-05-2019
Add to basket

Specifications

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]

Specifications

Copper foil
30 x 1000cm
7440-50-8
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
Odorless
23978
Foil
Undesignated
≈20.2g/30x30cm
0.025mm (0.001 in.) thick, annealed, uncoated
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
99.8%
Videos
SDS