missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate monohydrate, 98+%, extra pure, ACROS Organics™

Brand:  Acros Organics 203120025

Code : 29

Additional Details : CAS Number : 6046-93-1 Weight : 4.00000kg Transport : UN number : 3077 Chem class : 9 Pack group : III


Packaging Plastic bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 11944901

  • 181.00 € / 2.50kg

Estimated Shipment: 05-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Oxidation reagent
Specifications

Chemical Identifiers

6046-93-1
199.65
NWFNSTOSIVLCJA-UHFFFAOYSA-L
165397
O.[Cu++].CC([O-])=O.CC([O-])=O
C4H8CuO5
MFCD00149570
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
copper;diacetate;hydrate

Specifications

Copper(II) acetate monohydrate
98% min. (Iodometry)
(CH3CO2)2Cu·H2O
15,2614
Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether
O.[Cu++].CC([O-])=O.CC([O-])=O
199.65
199.65
98+%
30ppm max
1.88g/mL
0.01% max
Plastic bottle
100ppm max.
Blue to Blue-Green
2.5kg
6046-93-1
C4H8CuO5
MFCD00149570
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper;diacetate;hydrate
165397
Crystalline Powder
Extra Pure
30ppm max.
Authentic
20ppm max.
Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether
240.0°C
115.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Copper(II) acetate monohydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates