missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, 99+%, Thermo Scientific Chemicals

Catalog Number p-46619
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1317-38-0
CuO
79.55
MFCD00010979
KKCXRELNMOYFLS-UHFFFAOYSA-N
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
14829
[O--].[Cu++]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1317-38-0
CuO
79.55
MFCD00010979
KKCXRELNMOYFLS-UHFFFAOYSA-N
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
14829
[O--].[Cu++]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1326.0°C
Color Black
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula CuO
Fieser 04,108; 05,161
Merck Index 15, 2636
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dil.acids - slowly soluble in nh3, soluble in alkali cyanides and (nh4)2co3 solutions, practically insoluble in alcohol
IUPAC Name oxocopper
Formula Weight 79.54
Percent Purity ≥99%
Packaging Plastic bottle
Chemical Name or Material Copper(II) oxide, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Copper(II) oxide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'