missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Trifluoromethanesulfonate, 98%, ACROS Organics™

Brand:  Acros Organics 405420250

Code : 29

Additional Details : CAS Number : 34946-82-2 Weight : 0.07500kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10003933

  • 220.00 € / 25g

Estimated Shipment: 05-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

34946-82-2
361.69
GZWXEFRPSWBAGC-UHFFFAOYSA-N
45051791
C(F)(F)(F)S(=O)(=O)O.[Cu]
C2CuF6O6S2
MFCD00077492
copper; trifluoromethanesulfonic acid
copper;trifluoromethanesulfonic acid

Specifications

Copper(II) trifluoromethanesulfonate
Blue to White or Gray
34946-82-2
C2CuF6O6S2
MFCD00077492
copper; trifluoromethanesulfonic acid
C(F)(F)(F)S(=O)(=O)O.[Cu]
361.69
361.69
98%
Glass bottle
25g
98%
(CF3SO3)2Cu
07,76; 08,126; 10,110; 14,102
GZWXEFRPSWBAGC-UHFFFAOYSA-N
copper;trifluoromethanesulfonic acid
45051791
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Copper(II) trifluoromethanesulfonate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates