missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Copper/Plastic Funnel Filter
Specifically sized and suited for use in a funnel. Bochem™ Copper/Plastic Funnel Filter is made of two materials for durability and resistance.
6.75 € - 13.65 €
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Disc | Price | Quantity & Availability | |||||
10329199
|
Bochem™
8851 |
50mm |
6.75 €
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10058909
|
Bochem™
8853 |
105mm |
13.65 €
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- Copper and plastic construction.
- Resistant and durable.
- Comes in two sizes.
Specifications
Plastic | |
Funnel |