missing translation for 'onlineSavingsMsg'
Learn More

Copper powder, semispherical, -50+70 mesh, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number 15416685
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3077
Quantity 100 g
Percent Purity 99%
Mesh Size −50+70 Mesh
Odor Odorless
Assay (metals basis)
Chemical Name or Material Copper powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'