missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:Without Lid
Brand: Corning™ 3880
Code : 66
Additional Details : Weight : 0.03523kg
Description
Specifications
Half-Area | |
8 x 12 | |
Standard automated handling and detection systems | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
Without Lid | |
Black, Clear Bottom | |
Polystyrene | |
25 to 125 μL | |
Non-treated | |
Suitable for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface | |
25/Pk. | |
Clear |