missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap

Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.

Brand:  Corning™ 356485

Code : NEW

Additional Details : Weight : 4.06000kg

 View more versions of this product

Product Code. 10062501

  • 580.00 € / Pack of 50
Estimated Shipment: 20-09-2024
to see stock.
Add to basket

Description

Description

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • Specifications

    Specifications

    Yes
    Phenolic
    For adhesion, growth and differentiation of neuronal and transfected cell lines
    50/Cs.
    Rectangular
    Sterile
    Vented
    8.45 oz.
    250 mL
    Canted
    5/Pk.
    Collagen I
    Rectangular
    75 cm2
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers