Learn More
Corning™ BioCoat™ Intestinal Epithelium Differentiation Environment Kit
Used to create intestinal models that exhibit barrier function for use in transport and absorption studies of drugs, metabolites and infectious diseases of the intestine
Brand: Corning™ 355057
Code : NEW
Additional Details : Weight : 1.06000kg
Includes:
- 400mL Entero-STIM™ Enterocyte Differentiation Medium (DMEM containing sodium butyrate)
- Lyophilized BD™ MITO+ Serum Extender Supplement
- 100mL Seeding Basal Medium (DMEM)
- 24 ™ Fibrillar Collagen Cell Culture Inserts (inserts contain 1μm pore size membranes packaged in one 24-well Falcon™ Companion TC Plate with one additional plate
Description
Corning™ BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models. The system comprises a specially formulated serum-free medium, culture supplements, sodium butyrate, and Corning BioCoat Fibrillar Collagen Cell Culture Inserts in one convenient kit.
For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.
Specifications
Transport and absorption studies of drugs, metabolites and infectious diseases of the intestine | |
2° to 8°C | |
1/Pk. | |
1/Ea. |