missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix
Used for culture of epithelial, endothelial, and granulosa cell types under low serum conditions. Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix has low levels of endotoxin, and absence of contamination.
POA
Specifications
Content And Storage | 2° to 8°C |
---|---|
Concentration | 1 X |
Form | Liquid |
Product Type | DMEM and Ham's F-12, 50/50 Mix |
Sterility | Sterile |
Description
- Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
- Fifty-fifty mixture of DMEM and F-12, originally formulated for rat neuroblastoma cells and MDCK cells
- Serum-free growth may be possible with appropriate protein supplements
- Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
- Formulation of DMEM/Ham's F-12 contains L-glutamine
- Sterile
Specifications
2° to 8°C | |
Liquid | |
Sterile | |
To assess growth promotion and cytotoxicity characteristics |
1 X | |
DMEM and Ham's F-12, 50/50 Mix | |
<0.25EU/mL | |
7 to 7.4 |
IVD Product