Learn More
Corning™ Ciprofloxacin Hydrochloride
Packaged in plastic bottles
Brand: Corning™ 61-277-RG
Code : NEW
Additional Details : CAS Number : 1405-10-3 Weight : 0.09000kg
Specifications
15° to 30°C | |
Antibiotic | |
5 g |
Powder | |
Plastic Bottle |
Packaged in plastic bottles
Brand: Corning™ 61-277-RG
Code : NEW
Additional Details : CAS Number : 1405-10-3 Weight : 0.09000kg
15° to 30°C | |
Antibiotic | |
5 g |
Powder | |
Plastic Bottle |