Learn More
Corning™ Metallocene Collection Container 2D Pillow Configuration
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Brand:  Corning™ 91-100-35
Additional Details : Weight : 0.33770kg
 View more versions of this product
Product Code. 17213574
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Specifications
Specifications
Metallocene
100 L
1 Ea.
Sterile
Port 1, 2; 24 in. length tubing w/0.375 in. I.D. Male CPC conn. w/0.375 in. barb, dust cover and clamps; Port 3; 36 in. tubing 0.125 in. I.D. w/0.125 in. barb and male luer
1/Pk.