missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Culture Grade Water Tested to USP Sterile Water

High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.

Brand:  Corning™ 25-055-LG

Code : NEW

Additional Details : Weight : 1.00000kg

Quantity 1 x 100L

 View more versions of this product

Product Code. 15393651

  • 740.00 € / 100 L


Call for latest local availability
01 885 5854
Add to basket

Alert:

Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility


Description

Description

  • Tested to USP monograph for sterile water for injection (WFI)
  • Ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase, DNase, and Protease-free water is requiredFree of any added components
  • No toxic agents, including DEPC, are used, eliminating possible interferences with enzymatic reactions
  • Endotoxin levels ≤0.025EU/mL
  • Available in product sizes ranging from 100mL to 200L
Specifications

Chemical Identifiers

Specifications

1 x 100L
Videos