missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips, Nonsterile
Designed for quantitative assays in which sample binding can significantly alter results.
102.47 € - 329.17 €
Specifications
Sterility | Non-sterile |
---|---|
Color | Natural |
For Use With (Application) | Designed for quantitative assays in which sample binding can significantly alter results |
Format | Hinged Rack |
Tip Style | Low Retention, Round |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11951861
|
Corning™
4147 |
0.1 to 10 μL |
105.39 €
Pack of 960 |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||||
11971861
|
Corning™
4149 |
1 to 300 μL |
102.47 €
Pack of 960 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11961861
|
Corning™
4148 |
1 to 200 μL |
105.39 €
Pack of 960 |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||||
11323675
|
Corning™
4147 |
0.1 to 10 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11311132
|
Corning™
4148 |
1 to 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12999577
|
Corning™
4149 |
1 to 300 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Corning DeckWorks low binding tips are available in 10 μL, 200 μL, and 300 μL sizes.
- Proprietary low binding technology reduces surface tension on the interior wall of the tip.
- Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated.
- DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results.
- Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
Specifications
Non-sterile | |
Designed for quantitative assays in which sample binding can significantly alter results | |
Low Retention, Round | |
Universal Fit | |
DeckWorks |
Natural | |
Hinged Rack | |
Hinged rack | |
Yes | |
Polypropylene |