Learn More
Corning™ Disposable Vacuum Filter/Storage Systems
Features printed funnels that identify membrane type and product number for easy product product identification. Corning™ Disposable Vacuum Filter/Storage Systems provide a complete solution for your filtering needs. The system consists of a polystyrene filter funnel joined by an adapter ring to a removable polystyrene storage bottle with a separate sterile polyethylene cap.
128.60 € - 331.87 €
Specifications
Connectors | Angled hose |
---|---|
Description | Includes square membrane filter, graduated funnel and receiver of clear polystyrene, polyethylene neck adapter, orange HDPE plug seal |
Disposable | Single-use |
For Use With (Equipment) | 4.5cm bottle neck |
Material | Polystyrene |
Includes: Square membrane filter, graduated funnel and receiver of clear polystyrene; polyethylene neck adapter and plug cap (supplied sterile and individually packaged).
Product Code | Brand | Pore Size | Material (Membrane) | Volume (Metric) | Filtration Area | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Material (Membrane) | Volume (Metric) | Filtration Area | Price | Quantity & Availability | ||
11373785
|
Corning™
431475 |
0.1 μm | PES | 500 mL | 33.2 cm2 |
206.84 €
Pack of 12 |
In Stock
Log in to see stock. |
||
11363785
|
Corning™
431474 |
0.1 μm | PES | 1000 mL | 54.5 cm2 |
331.87 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10005341
|
Corning™
431154 |
0.2 μm | Cellulose Acetate | 150 mL | 13.6 cm2 |
128.60 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10468181
|
Corning™
430767 |
0.2 μm | Cellulose Acetate | 250 mL | 19.6 cm2 |
132.05 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10769922
|
Corning™
430769 |
0.2 μm | Cellulose Acetate | 500 mL | 33.2 cm2 |
216.88 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10768592
|
Corning™
431205 |
0.2 μm | Cellulose Acetate | 500 mL | 54.5 cm2 |
315.92 €
Pack of 12 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||
10689311
|
Corning™
430517 |
0.2 μm | Cellulose Acetate | 1000 mL | 54.5 cm2 |
315.92 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10104391
|
Corning™
430756 |
0.2 μm | Cellulose Nitrate | 250 mL | 19.6 cm2 |
132.05 €
Pack of 12 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||
10016110
|
Corning™
430758 |
0.2 μm | Cellulose Nitrate | 500 mL | 0.054 m2 |
216.88 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10377941
|
Corning™
430771 |
0.2 μm | Nylon | 250 mL | 19.6 cm2 |
132.05 €
Pack of 12 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||
10106961
|
Corning™
430773 |
0.2 μm | Nylon | 500 mL | 33.2 cm2 |
216.88 €
Pack of 12 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||
10660502
|
Corning™
430515 |
0.2 μm | Nylon | 1000 mL | 54.5 cm2 |
315.92 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10504211
|
Corning™
431153 |
0.2 μm | PES | 150 mL | 13.6 cm2 |
128.60 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10609141
|
Corning™
431096 |
0.2 μm | PES | 250 mL | 19.6 cm2 |
132.05 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10268951
|
Corning™
431097 |
0.2 μm | PES | 500 mL | 33.2 cm2 |
216.88 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10296191
|
Corning™
431098 |
0.2 μm | PES | 1000 mL | 54.5 cm2 |
315.92 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10164191
|
Corning™
430186 |
0.22 μm | Cellulose Nitrate | 1000 mL | 54.5 cm2 |
315.92 €
Pack of 12 |
In Stock
Log in to see stock. |
||
10174191
|
Corning™
431155 |
0.45 μm | Cellulose Acetate | 150 mL | 13.6 cm2 |
128.60 €
Pack of 12 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||
10350412
|
Corning™
430768 |
0.45 μm | Cellulose Acetate | 250 mL | 19.6 cm2 |
132.05 €
Pack of 12 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||
10229051
|
Corning™
430770 |
0.45 μm | Cellulose Acetate | 500 mL | 33.2 cm2 |
216.99 €
Pack of 12 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||
10075431
|
Corning™
431206 |
0.45 μm | Cellulose Acetate | 500 mL | 54.5 cm2 |
315.92 €
Pack of 12 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||
10730693
|
Corning™
430516 |
0.45 μm | Cellulose Acetate | 1000 mL | 54.5 cm2 |
315.92 €
Pack of 12 |
In Stock
Log in to see stock. |
||
Description
Receiver easily unscrews from unit and can be sealed with a plug cap for filtrate storage.
- Radiation sterilized and certified nonpyrogenic
- Angled hose connector simplifies vacuum line attachment
- Receiver bottles feature easy-grip sides for improved handling
- Units are color coded by membrane type and labeled with both membrane material and pore size
- All membrane types are tissue culture compatible, and heat-sealed to the support grid to maximize flowrate, and reduce foaming and protein denaturization
- NYL, CN and PES membranes are detergent-free
- Models with 150mL funnel contain 42 × 42mm square membrane filter
- 250mL funnel contains 49.5 × 49.5mm square membrane filter
- 500mL funnel contains 63 × 63mm square membrane filter
- 1000mL funnel contains 79 × 79mm square membrane filter
- Cellulose acetate (CA)—For low protein binding
- Nylon (NYL)—For low extractables
- Cellulose nitrate (CN)—For general-purpose filtration
- Polyethersulfone (PES)—For fast flowrates and low protein binding
Not for parenteral use.
Specifications
Angled hose | |
Single-use | |
Polystyrene | |
Yes | |
Vacuum Filter System |
Includes square membrane filter, graduated funnel and receiver of clear polystyrene, polyethylene neck adapter, orange HDPE plug seal | |
4.5cm bottle neck | |
Sterile | |
12 case |