Learn More
Corning™ Gentest™ Pre-Coated PAMPA Plate System
A 96-well permeable support system with a filter plate pre-coated with structured layers of phospholipids.
Brand: Corning™ 353015
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Stable, pre-coated PAMPA plate is highly reproducible
- Improved assay predictability over traditional PAMPA
- Reduced mass retention of compounds
- Well-suited as a ranking tool for the assessment of compounds with passive transport mechanisms and for predicting in vivo drug permeability
- Polystyrene construction