missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Membrane Matrix

Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.

Brand:  Corning™ 356235

Code : NEW

Additional Details : Weight : 0.15000kg

 View more versions of this product

Product Code. 10429212

  • 1627.05 € / 50mL


Estimated Shipment date
from Supplier 05-03-2019
Add to basket

Alert:

Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer


Description

Description

Solubilized basement membrane preparations polymerize at room temperature. 

  • Source: Engelbreth-Holm-Swarm (EHS) mouse tumor
  • Base formulation: Dulbecco's Modified Eagle's Medium with 50μg/mL gentamycin, with or without Phenol Red

 Phenol Red-free is available for assays that require color detection, including fluorescence assays

 Quality

  • Tested for the ability to promote neurite outgrowth of chick dorsal root ganglia cells
  • Tested and found negative for LDEV/LDHV, bacteria, fungi, and mycoplasma
  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Protein concentrations determined by Lowry method
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay
  • Matrigel Matrix gel stability tested for a period of 14 days at 37°C
  • Biological activity determined for each lot using a neurite outgrowth assay.
Specifications

Specifications

Cell growth and differentiation; metabolism/toxicology studies; invasion assays; in vitro and in vivo angiogenesis assays; in vivo angiogenesis studies and augmentation of tumors in immunosuppresssed mice
Basement membrane matrix
Engelbreth-Holm-Swarm Mouse Tumor
N/A
50mL
-20°C
Videos