missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Premium Fetal Bovine Serum

Used as a supplement to cell culture media to help sustain growth and metabolism

Brand:  Corning™ 35-016-CV

Code : NEW

Additional Details : Weight : 3.76000kg

 View more versions of this product

Product Code. 15393681

  • 622.00 € / 500mL


Call for latest local availability
01 885 5854
Add to basket

Description

Description

  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Premium FBS is sourced from U.S. herds
  • Heat-inactivated Fetal Bovine Serum (FBS) is heated at 56°C for 30 minutes
Specifications

Specifications

Sterile
cGMP
Yes
Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only.
315 ± 35mOsm/kg H20
30 to 50g/L
Triple 0.1μm Filtration
500mL
Heat Inactivated Premium FBS
Caramel/Amber
≤20EU/mL
Premium
<30mg/dL
7.8 ±0.8
-25° to -15°C
Bovine Fetus
USDA Inspected
Videos