Learn More
Corning™ Primaria™ Tissue Culture Flasks
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
Brand: Corning™ 353810
Code : NEW
Additional Details : Weight : 5.72400kg
Description
- Unique nitrogen-containing tissue culture surface chemistry
- Improves attachment, spreading, and growth for many primary cells or cell lines
- Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
- Stable, permanent surface modification
- Optically clear
- No special storage required
- Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
- Packaged in red color-coded, peel-open, medical-style packages
- Sterilized by gamma irradiation
- Nonpyrogenic tested to less than 0.1 EU/mL
Specifications
Blue | |
250 mL | |
Straight | |
5/Pk. | |
Sterile | |
Vented |
Phenolic | |
Polystyrene | |
100/Cs. | |
Rectangular | |
Rectangular | |
75 cm2 |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.