Learn More
Corning™ PureCoat™ Collagen I Peptide TC-Treated Microplates
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
Brand: Corning™ 356271
Code : NEW
Additional Details : Weight : 0.78000kg
Includes: Lid
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Specifications
cGMP | |
Yes | |
Flat | |
2 cm2 | |
Rectangular | |
Collagen Type I | |
24 | |
Lid | |
5/Pk. |
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Sterile | |
Cell Culture Microplate | |
Provides a highly consistent alternative to complex extracellular peptides | |
10/Cs. | |
Room Temperature |