Learn More
Corning™ PureCoat™ E cm Mimetic Multi-Flask
Features multi-layers with vented cap
Brand: Corning™ 356244
Code : NEW
Additional Details : Weight : 3.08000kg
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Compliance
cGMP manufacturing compliant (ISO 9001:2008, ISO 13485:2012)
Specifications
Blue | |
Individually packed in foil bag | |
8/Cs. | |
Rectangular | |
Sterile | |
Vented | |
Fibronectin Peptide |
Polypropylene | |
Straight | |
1/Pk. | |
PureCoat E cm Mimetic Fibronectin Peptid | |
Rectangular | |
525 cm2 |