missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PureCoat™ ECM Mimetic Multi-Flask

Features multi-layers with vented cap

Brand:  Corning™ 356245

Code : NEW

Additional Details : Weight : 3.33000kg

 View more versions of this product

Product Code. 15329830

  • 983.65 € / Pack of 6


Estimated Shipment date
from Supplier 27-07-2020
Add to basket

Description

Description

  • Animal component free, synthetic peptide coating covalently attached to cultureware
  • Compatible with wide range of serum-free, xeno-free, and animal-free media formulations
  • Also compatible with broad range of cell dissociation reagents
  • Streamlines cell culture workflow, reducing handling and contamination risk
  • Predictable scale-up and uniform cell growth
  • Non-cytotoxic
  • Sterile

Specifications

  • Flask shape: Rectangular
  • Neck Style: Straight    
  • Surface treatment: Fibronectin-coated
  • Cap material: Polypropylene
  • Available in 6/Pk. or 8/Pk.
  • Cell growth area: 525cm2 and 875cm2

Compliance

cGMP manufacturing compliant (ISO 9001:2008, ISO 13485:2012)

Specifications

Specifications

5-layer with blue vented PP cap
Individually packed in foil bag
Rectangular
875 cm2
Straight
Fibronectin Peptide
Videos
Documents