Learn More
Corning™ PureCoat™ Fibronectin Peptide TC-Treated Microplates
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
365.16 € - 397.60 €
Specifications
Certifications/Compliance | cGMP |
---|---|
Closure Color | Clear |
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Includes: Lid
Product Code | Brand | Well Count | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Count | Price | Quantity & Availability | |||||
15310001
|
Corning™
356240 |
6 |
365.16 €
Pack of 10 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
15330001
|
Corning™
356241 |
24 |
397.60 €
Pack of 10 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Specifications
cGMP | |
Yes | |
Flat | |
Polystyrene | |
Sterile | |
Cell Culture Microplate | |
Lid | |
1/Pk. |
Clear | |
Polystyrene | |
Clear | |
Rectangular | |
Fibronectin | |
Provides a highly consistent alternative to complex extracellular peptides | |
10/Cs. | |
Room Temperature |