Learn More
Corning™ Phenolic GPI Screw Caps for Disposable Culture Tubes
With white rubber liner
Brand:  Corning™ 99999-13
Code : 02
Additional Details : Weight : 1.90509kg
 View more versions of this product
Product Code. 10426843
  • 319.14 € / Pack of 1000
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
Made of a phenolic compound. These caps have a white rubber liner.
Specifications
Specifications
Screw Cap
Rubber
Phenolic
13.5 mm
1000/Cs.
13-415
Autoclavable
Black
1000/Pk.
Culture Tubes