missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ SF Medium
Based on a proprietary combination of classical media formulations
POA
Specifications
Content And Storage | 2° to 8°C |
---|---|
Sterility | Sterile |
pH | 7.2 to 7.6 |
Quantity | 6 x 500 mL |
Description
SF Medium is based on a proprietary classical media formulation. This medium contains a mixture of select trace elements and high molecular weight carbohydrates, extra vitamins, animal-free protein, and a small amount of high quality bovine serum albumin (1.0 g/L). SF Medium supports superior growth and viability over long-term passages in hybridomas, as well as suspension and adherent cell cultures. This formulation contains L-glutamine.
Specifications
2° to 8°C | |
Sterile | |
7.2 to 7.6 | |
6 x 500 mL |