Learn More
Corning™ SF Medium
Based on a proprietary combination of classical media formulations
Brand:  Corning™ 40-101-CV
Code : NEW
Additional Details : Weight : 5.44310kg
 View more versions of this product
Product Code. 15323701
  • 213.00 € / Pack of 6
EU Stock 6
Description
Description
  • Contains a mixture of select trace elements and high molecular carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1.0 g/L)
  • Supports superior growth and viability over long-term passages in hybridomas, as well as suspension and adherent cell cultures
  • Formulation contains L-glutamine
Offers a viable alternative to FBS for many hybridomas and works on a broad range of suspension and attachment cells; can be cell line dependent
Specifications
Specifications
7.2 to 7.6
6 x 500mL
Sterile
SF Medium, With L-Glutamine
2° to 8°C