Learn More
Emma Safety Footwear Corona Safety Shoes
Washable microfiber uppers are treated to repel all kinds of dirt, germs and bacteria. Highly breathable, comfortable shoe for all day wear. Energy returning soles prevent fatigue and keep the wearer fresh, alert, foot sure and steady even in the event of a spillage.
Brand:  Emma Safety Footwear MM54454039
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 15978721
  • 118.72 € / 1 pair
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • GripForce™ Easy Twist Outsole
  • Microfiber construction
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011
White
Hydro-Tec™ Sanitized Silver
39
Laces
Corona
Microfiber
Steel