missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Brand: Corning™ 3912
Code : 66
Additional Details : Weight : 6.19740kg
Description
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Non-treated | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. | |
25/Pk. |
Without Lid | |
White | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |