missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
441.92 € - 530.59 €
Specifications
Material | Polystyrene |
---|---|
No. of Wells | 96 |
Volume (Metric) Working | 75 to 200 μL |
Sterility | Non-sterile |
Well Shape | Flat Bottom (F-well) |
Product Code | Brand | Color | Sterility | Surface Treatment | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | Surface Treatment | Price | Quantity & Availability | |||
10679141
|
Corning™
3922 |
White | Non-sterile | High Binding |
492.07 €
Pack of 100 |
Estimated Shipment: 19-04-2024
Log in to see stock. |
|||
10133811
|
Corning™
3925 |
Black | Non-sterile | High Binding |
492.07 €
Pack of 100 |
Estimated Shipment: 19-04-2024
Log in to see stock. |
|||
10002551
|
Corning™
3912 |
White | Non-sterile | Non-treated |
441.92 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10022561
|
Corning™
3915 |
Black | Non-sterile | Non-treated |
530.59 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
Description
Specifications
Polystyrene | |
75 to 200 μL | |
Flat Bottom (F-well) |
96 | |
Non-sterile |