missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 314, 99%, pure, laser grade, ACROS Organics™

Brand:  Acros Organics 405620025

Code : 29

Additional Details : CAS Number : 55804-66-5 Weight : 1.50250kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Quantity 2.5g

 View more versions of this product

Product Code. 11956191

  • 698.00 € / 2.50g


Estimated Shipment: 07-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

55804-66-5
313.353
VMJKUPWQKZFFCX-UHFFFAOYSA-N
72653
CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3
C18H19NO4
MFCD00051334
1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one
CHEBI:51940

Specifications

Coumarin 314
99%
MFCD00051334
VMJKUPWQKZFFCX-UHFFFAOYSA-N
313.353
CHEBI:51940
99%
140.0°C to 144.0°C
55804-66-5
C18H19NO4
1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one
CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3
72653
313.34
Pure
2.5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Coumarin 314, laser grade

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates