Learn More
Hirschmann™ Cover Glasses for Microscopy
Preserve and protect specimens on slides. Hirschmann Instruments™ Cover Glasses for Microscopy are manufactured from quality glass and come in a range of dimensions.
Brand:  Hirschmann™ 8000111
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 13178159
  • 83.60 € / Pack of 1000
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
Description
Description
  • 0.13-0.17 mm thickness
  • Acc. to DIN 58 884, CE acc. IVD 98/79 EC
Specifications
Specifications
Coverslip
#1 (0.13 to 0.16 mm)
22 x 50 mm
CE, DIN 58884, IVD 98/79 EC